Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adam10
Entrez Gene ID
11487
Gene Name
a disintegrin and metallopeptidase domain 10
Gene Aliases
1700031C13Rik, MADM, kuz, kuzbanian
Chromosome Location
Chr.9: 70679001 - 70780229 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007399.3 NP_031425.2 8 - 9 1219 78
GenBank mRNA AF011379.1 - 8 - 9 1040 78
AK036599.1 - 2 - 3 263 78
BC057346.1 - 8 - 9 1180 78
BC066207.1 - 8 - 9 1224 78

View Full Product Details