The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024181.2 | NP_077143.2 | 9 - 10 | 1338 | 58 |
GenBank mRNA | AF255459.1 | - | 9 - 10 | 1015 | 58 |
AF314002.1 | - | 8 - 9 | 851 | 58 | |
AK004617.1 | - | 9 - 10 | 1093 | 58 | |
AK028184.1 | - | 9 - 10 | 1078 | 58 | |
AK128945.1 | - | 9 - 10 | 1092 | 58 | |
AK165652.1 | - | 9 - 10 | 1097 | 58 | |
AK169020.1 | - | 9 - 10 | 1105 | 58 |