Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Poteg
Entrez Gene ID
70952
Gene Name
POTE ankyrin domain family, member G
Gene Aliases
4921537P18Rik, 4930456F22Rik, Gsarp1, Gsarp2
Chromosome Location
Chr.8: 27447529 - 27582715 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026256.2 NP_080532.1 1 - 2 390 65
NM_027618.1 NP_081894.1 1 - 2 390 65
GenBank mRNA AK015012.1 - 1 - 2 391 65
AK015165.1 - 1 - 2 391 65
AK015473.1 - 1 - 2 391 65
DQ298444.1 - 1 - 2 391 65
DQ517440.1 - 1 - 2 391 65

View Full Product Details