Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ap1s2
Entrez Gene ID
108012
Gene Name
adaptor-related protein complex 1, sigma 2 subunit
Gene Aliases
1500012A13Rik, AI853648, EST1
Chromosome Location
Chr.X: 163907377 - 163933666 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290378.1 NP_001277307.1 1 - 2 257 98
NM_001290379.1 NP_001277308.1 1 - 2 257 98
NM_026887.3 NP_081163.2 1 - 2 257 98
GenBank mRNA AK043776.1 - 1 - 2 78 98
AK045558.1 - 1 - 2 258 98
AK047724.1 - 1 - 2 134 98
AK053941.1 - 1 - 2 130 98
AK167690.1 - 1 - 2 132 98
BC046964.1 - 1 - 2 114 98

View Full Product Details