Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stk11ip
Entrez Gene ID
71728
Gene Name
serine/threonine kinase 11 interacting protein
Gene Aliases
1200014D22Rik, BB131189, LIP1, LKB1IP
Chromosome Location
Chr.1: 75521529 - 75537335 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027886.3 NP_082162.3 6 - 7 688 75
GenBank mRNA AK004757.2 - 6 - 7 641 75
AK156122.1 - 5 - 6 776 75
AK165911.1 - 6 - 7 679 75
AK171989.1 - 5 - 6 550 75
BC042403.1 - 5 - 6 547 75
BC061635.1 - 6 - 7 671 75

View Full Product Details