The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027903.3 | NP_082179.1 | 5 - 6 | 775 | 75 |
GenBank mRNA | AK005050.2 | - | 5 - 6 | 776 | 75 |
AK154452.1 | - | 5 - 6 | 757 | 75 | |
BC031710.1 | - | 5 - 6 | 753 | 75 | |
BC116414.2 | - | 5 - 6 | 759 | 75 |