Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psd
Entrez Gene ID
73728
Gene Name
pleckstrin and Sec7 domain containing
Gene Aliases
1110007H17Rik, A930015K15, Efa6, Efa6a, Psdl, mKIAA2011
Chromosome Location
Chr.19: 46312085 - 46327184 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028627.2 NP_082903.2 17 - 18 3054 65
GenBank mRNA AK003542.1 - 1 - 2 29 65
AK158685.1 - 7 - 8 860 65
AK158851.1 - 7 - 8 986 65
AK220438.1 - 12 - 13 3171 65
BC043135.1 - 13 - 14 1883 65
BC057353.1 - 17 - 18 3055 65
BC058352.1 - 3 - 4 429 65
BC138652.1 - 15 - 16 2913 65
BC145352.1 - 15 - 16 2916 65

View Full Product Details