The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164609.1 | NP_001158081.1 | 12 - 13 | 1230 | 61 |
NM_001164610.1 | NP_001158082.1 | 12 - 13 | 1230 | 61 | |
NM_001164611.1 | NP_001158083.1 | 12 - 13 | 1230 | 61 | |
NM_029945.3 | NP_084221.2 | 13 - 14 | 1317 | 61 | |
GenBank mRNA | AK019466.2 | - | 13 - 14 | 1318 | 61 |
AK031029.1 | - | 12 - 13 | 1224 | 61 | |
AK031685.1 | - | 12 - 13 | 1229 | 61 | |
AK035892.1 | - | 12 - 13 | 1222 | 61 | |
AK036055.1 | - | 12 - 13 | 1222 | 61 | |
AK036380.1 | - | 12 - 13 | 1226 | 61 | |
AK129354.1 | - | 13 - 14 | 1294 | 61 | |
AK138720.1 | - | 12 - 13 | 1204 | 61 | |
AK164234.1 | - | 12 - 13 | 1222 | 61 | |
AK164456.1 | - | 10 - 11 | 1695 | 61 | |
BC026767.1 | - | 12 - 13 | 1203 | 61 |