The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164609.1 | NP_001158081.1 | 15 - 16 | 1531 | 53 |
NM_001164610.1 | NP_001158082.1 | 14 - 15 | 1492 | 53 | |
NM_001164611.1 | NP_001158083.1 | 14 - 15 | 1489 | 53 | |
NM_029945.3 | NP_084221.2 | 15 - 16 | 1579 | 53 | |
GenBank mRNA | AK019466.2 | - | 15 - 16 | 1580 | 53 |
AK031029.1 | - | 14 - 15 | 1483 | 53 | |
AK031685.1 | - | 14 - 15 | 1491 | 53 | |
AK035892.1 | - | 14 - 15 | 1484 | 53 | |
AK036055.1 | - | 14 - 15 | 1484 | 53 | |
AK036380.1 | - | 14 - 15 | 1488 | 53 | |
AK129354.1 | - | 15 - 16 | 1556 | 53 | |
AK138720.1 | - | 14 - 15 | 1466 | 53 | |
AK164234.1 | - | 15 - 16 | 1523 | 53 | |
AK164456.1 | - | 12 - 13 | 1957 | 53 | |
BC026767.1 | - | 14 - 15 | 1465 | 53 |