The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053178.2 | NP_444408.1 | 1 - 2 | 334 | 100 |
GenBank mRNA | AB050554.1 | - | 1 - 2 | 334 | 100 |
AK054103.1 | - | 1 - 2 | 147 | 100 | |
AK129179.1 | - | 1 - 2 | 130 | 100 | |
BC057322.1 | - | 1 - 2 | 130 | 100 |