The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133796.7 | NP_598557.3 | 3 - 4 | 393 | 103 |
GenBank mRNA | AK075656.1 | - | 2 - 3 | 260 | 103 |
AK077343.1 | - | 3 - 4 | 553 | 103 | |
AK143516.1 | - | 3 - 4 | 377 | 103 | |
AK147842.1 | - | - | 352 | 103 | |
AK156819.1 | - | - | 338 | 103 | |
AK165241.1 | - | 3 - 4 | 375 | 103 | |
AK170553.1 | - | 3 - 4 | 360 | 103 | |
AK170718.1 | - | 3 - 4 | 377 | 103 | |
AK172140.1 | - | - | 338 | 103 | |
BC004732.1 | - | 3 - 4 | 335 | 103 | |
BC086755.1 | - | 3 - 4 | 317 | 103 |