Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dner
Entrez Gene ID
227325
Gene Name
delta/notch-like EGF-related receptor
Gene Aliases
A930026D19Rik, BET, Bret
Chromosome Location
Chr.1: 84369839 - 84696221 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152915.1 NP_690879.1 5 - 6 1485 83
GenBank mRNA AB067650.1 - 5 - 6 1134 83
AF370126.1 - 5 - 6 1141 83
AK044597.1 - 5 - 6 1248 83
AY032924.1 - 5 - 6 1141 83
BC034634.1 - 5 - 6 1485 83

View Full Product Details