The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152915.1 | NP_690879.1 | 9 - 10 | 2102 | 92 |
GenBank mRNA | AB067650.1 | - | 9 - 10 | 1751 | 92 |
AF370126.1 | - | 9 - 10 | 1758 | 92 | |
AK044597.1 | - | 9 - 10 | 1865 | 92 | |
AK152755.1 | - | 4 - 5 | 505 | 92 | |
AK153235.1 | - | 4 - 5 | 505 | 92 | |
AY032924.1 | - | 9 - 10 | 1758 | 92 | |
BC022636.1 | - | 4 - 5 | 565 | 92 | |
BC034634.1 | - | 9 - 10 | 2102 | 92 |