Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kdm4b
Entrez Gene ID
193796
Gene Name
lysine (K)-specific demethylase 4B
Gene Aliases
4732474L06Rik, Jmjd2b, mKIAA0876
Chromosome Location
Chr.17: 56326033 - 56403769 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172132.2 NP_742144.1 9 - 10 1318 62
GenBank mRNA AK078074.1 - 9 - 10 1311 62
AK129233.1 - 9 - 10 1277 62
AK141879.1 - 9 - 10 1295 62
AK145151.1 - 9 - 10 1270 62
BC007145.1 - 9 - 10 1288 62

View Full Product Details