The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163073.1 | NP_001156545.1 | 6 - 7 | 817 | 93 |
NM_172153.3 | NP_742165.1 | 6 - 7 | 830 | 93 | |
GenBank mRNA | AB076078.1 | - | 6 - 7 | 829 | 93 |
AK041987.1 | - | 6 - 7 | 776 | 93 | |
AK154398.1 | - | 6 - 7 | 778 | 93 | |
AK155421.1 | - | 6 - 7 | 778 | 93 |