Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dync2li1
Entrez Gene ID
213575
Gene Name
dynein cytoplasmic 2 light intermediate chain 1
Gene Aliases
4933404O11Rik, CGI-60, D2lic, LIC3, mD2LIC
Chromosome Location
Chr.17: 84626496 - 84655568 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172256.1 NP_758460.1 10 - 11 889 76
GenBank mRNA AK008822.1 - 9 - 10 839 76
AK160259.1 - 10 - 11 903 76
BC030450.1 - 10 - 11 891 76
BC039070.1 - 10 - 11 889 76

View Full Product Details