The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172279.1 | NP_758483.1 | 14 - 15 | 1596 | 140 |
GenBank mRNA | AK035668.1 | - | 2 - 3 | 264 | 140 |
AK122565.2 | - | 10 - 11 | 1242 | 140 | |
AK146784.1 | - | 15 - 16 | 1931 | 140 | |
AY151083.1 | - | 14 - 15 | 1596 | 140 |