The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172295.4 | NP_758499.3 | 6 - 7 | 1379 | 93 |
GenBank mRNA | AK029040.1 | - | 6 - 7 | 1118 | 93 |
AK161057.1 | - | 6 - 7 | 1296 | 93 | |
AK166928.1 | - | - | 1129 | 93 | |
AK172389.1 | - | 6 - 7 | 1328 | 93 | |
BC037703.1 | - | 6 - 7 | 1316 | 93 |