Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Celf3
Entrez Gene ID
78784
Gene Name
CUGBP, Elav-like family member 3
Gene Aliases
4930415M08Rik, BRUNOL1, CAGH4, ERDA4, Tnrc4
Chromosome Location
Chr.3: 94478285 - 94492199 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289613.1 NP_001276542.1 4 - 5 959 85
NM_001289616.1 NP_001276545.1 5 - 6 441 85
NM_001289620.1 NP_001276549.1 5 - 6 441 85
NM_172434.3 NP_766022.1 4 - 5 956 85
NR_110353.1 3 - 4 267 85
GenBank mRNA AK019578.1 - 2 - 3 668 85
AK030259.1 - 3 - 4 267 85
AK044069.1 - 4 - 5 389 85
AK044506.1 - 5 - 6 405 85
AK087536.1 - 5 - 6 401 85
AY165052.1 - 4 - 5 674 85
BC031740.1 - 1 - 2 120 85
BC057553.1 - 4 - 5 1014 85

View Full Product Details