The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289613.1 | NP_001276542.1 | 7 - 8 | 1331 | 79 |
NM_001289616.1 | NP_001276545.1 | 8 - 9 | 813 | 79 | |
NM_001289620.1 | NP_001276549.1 | 8 - 9 | 813 | 79 | |
NM_172434.3 | NP_766022.1 | 7 - 8 | 1328 | 79 | |
NR_110353.1 | 6 - 7 | 639 | 79 | ||
GenBank mRNA | AK030259.1 | - | 6 - 7 | 639 | 79 |
AK044069.1 | - | 7 - 8 | 761 | 79 | |
AK044506.1 | - | 8 - 9 | 777 | 79 | |
AK087536.1 | - | 8 - 9 | 775 | 79 | |
AY165052.1 | - | 7 - 8 | 1046 | 79 | |
BC031740.1 | - | 4 - 5 | 492 | 79 | |
BC057553.1 | - | 7 - 8 | 1386 | 79 |