The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172449.2 | NP_766037.2 | 23 - 24 | 5666 | 70 |
GenBank mRNA | AK051895.1 | - | 2 - 3 | 1615 | 70 |
AK122329.1 | - | 1 - 2 | 537 | 70 | |
BC055691.1 | - | 3 - 4 | 417 | 70 | |
BC066146.1 | - | 2 - 3 | 677 | 70 | |
BC083181.1 | - | 5 - 6 | 640 | 70 | |
EL605892.1 | - | 1 - 2 | 70 |