The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172453.3 | NP_766041.1 | 10 - 11 | 2308 | 66 |
GenBank mRNA | AK049353.1 | - | 10 - 11 | 2310 | 66 |
AK154072.1 | - | 10 - 11 | 2354 | 66 | |
AY100322.1 | - | 11 - 12 | 2021 | 66 | |
AY498715.1 | - | 10 - 11 | 2308 | 66 | |
BC024481.1 | - | 4 - 5 | 442 | 66 | |
BC046611.1 | - | 11 - 12 | 1946 | 66 | |
BC060108.1 | - | 8 - 9 | 2519 | 66 |