The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010593.2 | NP_034723.1 | 1 - 2 | 153 | 77 |
GenBank mRNA | AK145895.1 | - | 1 - 2 | 138 | 77 |
AK170934.1 | - | 1 - 2 | 126 | 77 | |
AK171501.1 | - | - | 127 | 77 | |
BC040757.1 | - | 1 - 2 | 102 | 77 | |
BC094461.1 | - | 1 - 2 | 106 | 77 | |
CD555608.1 | - | 1 - 2 | 153 | 77 |