Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Jup
Entrez Gene ID
16480
Gene Name
junction plakoglobin
Gene Aliases
Ctnng, D930025P04Rik, PG
Chromosome Location
Chr.11: 100368856 - 100397790 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010593.2 NP_034723.1 2 - 3 373 82
GenBank mRNA AK145895.1 - 2 - 3 358 82
AK170934.1 - 2 - 3 346 82
AK171501.1 - - 347 82
BC040757.1 - 2 - 3 322 82
BC094461.1 - 2 - 3 326 82
CD555608.1 - 2 - 3 373 82

View Full Product Details