Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Jup
Entrez Gene ID
16480
Gene Name
junction plakoglobin
Gene Aliases
Ctnng, D930025P04Rik, PG
Chromosome Location
Chr.11: 100368856 - 100397790 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010593.2 NP_034723.1 13 - 14 2418 84
GenBank mRNA AK086401.1 - 1 - 2 429 85
AK145895.1 - 13 - 14 2403 84
AK170934.1 - 13 - 14 2391 84
BC040757.1 - 13 - 14 2367 84
BC094461.1 - 13 - 14 2371 84

View Full Product Details