Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpt1a
Entrez Gene ID
12894
Gene Name
carnitine palmitoyltransferase 1a, liver
Gene Aliases
C730027G07, CPTI, Cpt1
Chromosome Location
Chr.19: 3322326 - 3385735 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013495.2 NP_038523.2 14 - 15 1828 93
GenBank mRNA AF017175.1 - - 1716 93
AK077454.1 - 14 - 15 1782 93
AK128925.1 - 14 - 15 1782 93
AK133472.1 - 14 - 15 1783 93
AK136487.1 - 14 - 15 1782 93
AK147770.1 - 14 - 15 1783 93
AK149688.1 - 14 - 15 1779 93
BC038395.1 - 13 - 14 1763 93
BC046383.1 - 13 - 14 1763 93
BC054791.1 - 14 - 15 1809 93
D49472.1 - 13 - 14 1755 93

View Full Product Details