The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172493.2 | NP_766081.1 | 8 - 9 | 1119 | 124 |
GenBank mRNA | AB713187.1 | - | 9 - 10 | 914 | 124 |
AK088648.1 | - | 8 - 9 | 1053 | 124 | |
AK153986.1 | - | 8 - 9 | 1107 | 124 | |
AY312280.1 | - | 8 - 9 | 1056 | 124 |