The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027088.2 | NP_081364.1 | 3 - 4 | 250 | 82 |
GenBank mRNA | AB047820.1 | - | 3 - 4 | 151 | 82 |
AK079374.1 | - | 3 - 4 | 266 | 82 | |
AK129108.1 | - | 3 - 4 | 158 | 82 | |
AK170576.1 | - | 3 - 4 | 251 | 82 | |
AK171561.1 | - | 3 - 4 | 252 | 82 | |
BC042489.1 | - | 3 - 4 | 201 | 82 | |
BC050901.1 | - | 3 - 4 | 213 | 82 |