Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mipep
Entrez Gene ID
70478
Gene Name
mitochondrial intermediate peptidase
Gene Aliases
5730405E07Rik, MIP
Chromosome Location
Chr.14: 60784562 - 60905481 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027436.3 NP_081712.2 3 - 4 562 67
NR_040642.1 3 - 4 562 67
GenBank mRNA AK017491.1 - 3 - 4 538 67
AK038479.1 - 3 - 4 556 67
AK044460.1 - 3 - 4 547 67
AK049000.1 - 3 - 4 503 67
AK076613.1 - 3 - 4 490 67
AK146428.1 - 4 - 5 553 67
BC110358.1 - 3 - 4 540 67
BC144798.1 - 3 - 4 543 67
BC145711.1 - 3 - 4 543 67
CA460475.1 - 3 - 4 599 67

View Full Product Details