The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027436.3 | NP_081712.2 | 4 - 5 | 653 | 130 |
NR_040642.1 | 4 - 5 | 653 | 130 | ||
GenBank mRNA | AK017491.1 | - | 4 - 5 | 629 | 130 |
AK038479.1 | - | 4 - 5 | 647 | 130 | |
AK044460.1 | - | 4 - 5 | 638 | 130 | |
AK076613.1 | - | 4 - 5 | 581 | 130 | |
AK146428.1 | - | 5 - 6 | 644 | 130 | |
BC110358.1 | - | 4 - 5 | 631 | 130 | |
BC144798.1 | - | 4 - 5 | 634 | 130 | |
BC145711.1 | - | 4 - 5 | 634 | 130 |