The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027629.3 | NP_081905.1 | 12 - 13 | 1951 | 90 |
GenBank mRNA | AK016437.1 | - | 10 - 11 | 1310 | 90 |
AK039189.1 | - | 12 - 13 | 1935 | 90 | |
AK040513.1 | - | 11 - 12 | 1681 | 90 | |
BC020351.1 | - | 12 - 13 | 2164 | 90 | |
BC032150.1 | - | 8 - 9 | 1157 | 90 | |
BC076571.1 | - | 12 - 13 | 1848 | 90 | |
BG071667.2 | - | 2 - 3 | 90 |