Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccser2
Entrez Gene ID
72972
Gene Name
coiled-coil serine rich 2
Gene Aliases
1700012P13Rik, 2900054P12Rik, AI035535, AW108503, Fam190b, Gcap14
Chromosome Location
Chr.14: 36874936 - 36968764 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028407.3 NP_082683.2 1 - 2 38 68
GenBank mRNA AK005931.1 - 1 - 2 38 68
BC048641.1 - 1 - 2 37 68
BC060947.1 - 1 - 2 37 68

View Full Product Details