The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029362.3 | NP_083638.1 | 2 - 3 | 519 | 68 |
GenBank mRNA | AK008205.1 | - | 2 - 3 | 484 | 68 |
AK156473.1 | - | 2 - 3 | 479 | 68 | |
AK159193.1 | - | 2 - 3 | 481 | 68 | |
AK164550.1 | - | - | 408 | 68 | |
BC006905.1 | - | 2 - 3 | 235 | 68 | |
BC011429.1 | - | 2 - 3 | 496 | 68 | |
BC059279.1 | - | 2 - 3 | 471 | 68 |