The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163470.1 | NP_001156942.1 | 6 - 7 | 960 | 60 |
NM_172275.2 | NP_758479.2 | 6 - 7 | 960 | 60 | |
GenBank mRNA | AK150038.1 | - | 6 - 7 | 929 | 60 |
BC023072.1 | - | 2 - 3 | 343 | 60 | |
BC038396.1 | - | 6 - 7 | 916 | 60 |