The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001326.1 | NP_001001326.1 | 5 - 6 | 1738 | 62 |
NM_029811.2 | NP_084087.2 | 4 - 5 | 487 | 62 | |
NR_037617.1 | 5 - 6 | 1919 | 62 | ||
GenBank mRNA | AK158270.1 | - | 5 - 6 | 1743 | 62 |
BC030391.1 | - | 1 - 2 | 213 | 62 | |
BC051034.1 | - | 5 - 6 | 1919 | 62 | |
CF737707.1 | - | 2 - 3 | 220 | 62 |