The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001254762.1 | NP_001241691.1 | 1 - 2 | 433 | 78 |
NM_030037.2 | NP_084313.1 | 2 - 3 | 586 | 78 | |
GenBank mRNA | AK088731.1 | - | 1 - 2 | 424 | 78 |
BC003880.1 | - | 1 - 2 | 443 | 78 | |
BC034167.1 | - | 2 - 3 | 571 | 78 | |
BC039620.1 | - | 2 - 3 | 567 | 78 | |
BE861793.1 | - | 1 - 2 | 60 | 78 | |
CK617131.1 | - | 1 - 2 | 384 | 78 |