The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030219.2 | NP_084495.2 | 1 - 2 | 459 | 79 |
GenBank mRNA | AK019654.1 | - | 1 - 2 | 460 | 79 |