The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053084.2 | NP_444314.2 | 1 - 2 | 80 | 60 |
GenBank mRNA | AF347694.1 | - | 1 - 2 | 48 | 60 |
AK154028.1 | - | 1 - 2 | 45 | 60 | |
BY280460.1 | - | 1 - 2 | 80 | 60 |