The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138677.2 | NP_619618.1 | 11 - 12 | 1972 | 84 |
GenBank mRNA | AB042828.1 | - | 11 - 12 | 1961 | 84 |
AK030943.1 | - | 11 - 12 | 2192 | 84 | |
AK037889.1 | - | 11 - 12 | 1535 | 84 | |
AK082920.1 | - | 11 - 12 | 1925 | 84 | |
AK129090.1 | - | 11 - 12 | 1939 | 84 | |
AK135969.1 | - | 6 - 7 | 836 | 84 | |
AK151668.1 | - | 11 - 12 | 1947 | 84 | |
AK152724.1 | - | 11 - 12 | 1948 | 84 | |
AK152979.1 | - | 11 - 12 | 1944 | 84 | |
BC023237.1 | - | 11 - 12 | 1920 | 84 |