The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144526.3 | NP_653109.2 | 1 - 2 | 36 | 61 |
GenBank mRNA | AK049710.1 | - | 1 - 2 | 37 | 61 |
AK078429.1 | - | 1 - 2 | 37 | 61 | |
BC029115.1 | - | 1 - 2 | 28 | 61 |