The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172287.2 | NP_758491.1 | 2 - 3 | 351 | 75 |
GenBank mRNA | BC049152.1 | - | 2 - 3 | 351 | 75 |
BC054072.1 | - | 2 - 3 | 351 | 75 |