Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tarsl2
Entrez Gene ID
272396
Gene Name
threonyl-tRNA synthetase-like 2
Gene Aliases
A530046H20Rik
Chromosome Location
Chr.7: 65644898 - 65692093 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172310.2 NP_758514.2 5 - 6 818 100
GenBank mRNA AK040927.1 - 5 - 6 791 100
AK041280.1 - 5 - 6 818 100
AK044290.1 - 5 - 6 819 100
AK079416.1 - 5 - 6 801 100
BC039225.1 - 5 - 6 802 100

View Full Product Details