The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290419.1 | NP_001277348.1 | 23 - 24 | 4240 | 71 |
NM_172339.4 | NP_758842.1 | 22 - 23 | 4121 | 71 | |
GenBank mRNA | AK077522.1 | - | 22 - 23 | 4088 | 71 |
BC044754.2 | - | 7 - 8 | 2538 | 71 | |
BC057031.1 | - | 23 - 24 | 4206 | 71 |