The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172503.3 | NP_766091.2 | 10 - 11 | 2090 | 128 |
GenBank mRNA | AK052168.1 | - | 10 - 11 | 2104 | 128 |
AK053663.1 | - | 10 - 11 | 2104 | 128 | |
AK131119.1 | - | 5 - 6 | 948 | 128 | |
AK150221.1 | - | 10 - 11 | 2105 | 128 | |
AK155465.1 | - | 10 - 11 | 2104 | 128 | |
BC058080.1 | - | 10 - 11 | 2090 | 128 |