Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dis3l
Entrez Gene ID
213550
Gene Name
DIS3 mitotic control homolog (S. cerevisiae)-like
Gene Aliases
AV340375
Chromosome Location
Chr.9: 64306756 - 64341423 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001295.2 NP_001001295.2 12 - 13 2331 84
NM_001177784.1 NP_001171255.1 12 - 13 2292 84
NM_172519.3 NP_766107.1 12 - 13 2235 84
GenBank mRNA AK029974.1 - 12 - 13 2236 84
AK032451.1 - 12 - 13 2239 84
AK077031.1 - 12 - 13 2295 84
AK150851.1 - 12 - 13 2277 84
AK151780.1 - 12 - 13 2277 84
AK153433.1 - 12 - 13 2277 84
AK173310.1 - 12 - 13 2265 84
BC056939.1 - 12 - 13 2264 84

View Full Product Details