The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172526.3 | NP_766114.1 | 3 - 4 | 936 | 86 |
GenBank mRNA | AK039884.1 | - | 3 - 4 | 937 | 86 |
BC147554.1 | - | 2 - 3 | 640 | 86 | |
BC147555.1 | - | 2 - 3 | 640 | 86 |