Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ttc24
Entrez Gene ID
214191
Gene Name
tetratricopeptide repeat domain 24
Gene Aliases
A430025D11Rik
Chromosome Location
Chr.3: 88069410 - 88081987 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172526.3 NP_766114.1 5 - 6 1177 78
GenBank mRNA AK039884.1 - 5 - 6 1178 78
BC147554.1 - 4 - 5 881 78
BC147555.1 - 4 - 5 881 78

View Full Product Details