The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172551.3 | NP_766139.2 | 2 - 3 | 216 | 118 |
GenBank mRNA | AK030039.1 | - | 2 - 3 | 219 | 118 |
AK047545.1 | - | 2 - 3 | 209 | 118 | |
AK053356.1 | - | 2 - 3 | 207 | 118 | |
AK085666.1 | - | 2 - 3 | 209 | 118 | |
AK128956.1 | - | 2 - 3 | 208 | 118 | |
AK154181.1 | - | 2 - 3 | 207 | 118 | |
AK154729.1 | - | 2 - 3 | 198 | 118 | |
BC110697.1 | - | 2 - 3 | 192 | 118 | |
BC117939.1 | - | 2 - 3 | 209 | 118 | |
BC117940.1 | - | 2 - 3 | 209 | 118 |