The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172553.4 | NP_766141.1 | 2 - 3 | 701 | 76 |
GenBank mRNA | AK047846.1 | - | 2 - 3 | 702 | 76 |
AK086731.1 | - | 2 - 3 | 514 | 76 | |
BC052200.1 | - | 2 - 3 | 683 | 76 |