Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zdhhc17
Entrez Gene ID
320150
Gene Name
zinc finger, DHHC domain containing 17
Gene Aliases
A230053P19Rik, BB187739, D130071N24Rik, Hip14
Chromosome Location
Chr.10: 110941779 - 111010066 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172554.2 NP_766142.2 13 - 14 1460 106
GenBank mRNA AK039575.1 - 13 - 14 1461 106
AK046891.1 - 13 - 14 1402 106
AK122406.1 - 12 - 13 2405 106
BC027101.1 - 13 - 14 1348 106
BC117788.1 - 13 - 14 1402 106
BC117789.2 - 13 - 14 1402 106

View Full Product Details